BRAND® nano-cap™ Pipette Tips, BrandTech®
Supplier: Brandtech
732022 732102 732202 732222 732002
10033-972PK 1186.56 CAD
10033-972 10033-984 10033-996 10034-016 10034-028 10034-126
BRAND® nano-cap™ Pipette Tips, BrandTech®
Pipette Tips
BRAND nano-cap pipette tips are a shorter style (37 mm) ultra-micro/crystal tip for use on many pipettes with a nominal/maximum volume less than 20 µl.
  • Short (37 mm) ultramicro/"crystal" tips
  • Fits most major brands of pipettes
  • Suitable for Transferpette® sub-micro (0.1 - 1 µl) pipettes
All tips are produced under cleanroom conditions. Bulk tips are packed in resealable bags. New environmentally friendly refillable TipBox design is rigid and allows partial loading of multichannels. Hinged lid can be rotated 180 degrees to be a lift-off design.More Product Information