Blood Bank Cross Match Holders, Dynalon
Supplier: Dynalon
159565-4 159565-2 159565-1
71070-080CS 1801.37 CAD
71070-080 71070-084 71070-086
Blood Bank Cross Match Holders, Dynalon
Racks Tube Racks
Blood bank cross match holders are manufactured of clear acrylic shelves with white bases and sides
For typing, screening and other blood bank applications. For blood donor processing. Holds donor sample tubes as well as tubes for donor's serum and cell suspension, ABO, Rh, antibody grouping and screening. No on rack labeling supplied.
Ordering information: Additional sizes and materials are available upon request.