Thick Chromatography Paper, Grade 238, Ahlstrom
Supplier: Ahlstrom
2388-2020 2388-4657 2388-1015 2388-1416 2388-1515 2388-0710 2388-1618 2388-0575 2388-0708 2388-1114 2388-1214 2388-1417 2388-1517 2388-1616 2388-1520 2388-1834 2388-2025 2388-3343 2388-3545 2388-5252 2388-5868 2381-1417 238-25X30CM
28334-178PK 217.27 CAD
28334-178 28342-036 28342-038 28342-040 28342-042 47745-886 47745-888 47749-816 47749-818 47749-820 47749-822 47749-824 47749-826 47749-828 47749-830 47749-832 47749-834 47749-836 47749-838 47749-840 47749-842 490005-068 490005-070
Thick Chromatography Paper, Grade 238, Ahlstrom
Blotting Papers
Ahlstrom Grade 238 is a medium thick 100% cotton chromatography paper recommended for general applications
It is also ideal for use as wicks in protein or nucleic acid blotting.