Thick Chromatography Paper, Grade 237, Ahlstrom
Supplier: Ahlstrom
2378-0710 2378-1015 2378-2020 2378-4657 2378-0708
47745-890PK 163.44 CAD
47745-890 47745-892 47745-894 47745-896 490005-066
Thick Chromatography Paper, Grade 237, Ahlstrom
Blotting Papers
Ahlstrom Grade 237 is a medium thick 100% cotton chromatography paper recommended for general applications with medium-heavy solute loading
It is also ideal for use as wicks in protein or nucleic acid blotting.