Spatula Scrapers, Dynalon
Supplier: Dynalon
490864 490834
13450-714EA 289.85 CAD
13450-714 15551-282
Spatula Scrapers, Dynalon
Scrapers
A short handled unit specially designed as a scraper and spatula
Scrape and clean any hard to clean containers. 13 1/2" overall length. Polyethylene blade is 4 1/2" wide, 7" long. Complete with large hand-grip.