Open-Top Polypropylene Screw Thread Caps, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
73806A-24400 73806-24400 73806A-18400 73806A-20400
66014-380CS 73.91 CAD
66014-380 66009-967 66014-397 66014-398
Open-Top Polypropylene Screw Thread Caps, Kimble Chase
Closures Test/Sample Tube Closures
Economical, one-piece construction caps are autoclavable and designed from ASTM Specification E982
Caps are open-top and do not come with a septa.