Histology Freeze Plate, PolyScience

Supplier: Polyscience
HPLAT-30-A12E
10066-630EA 4849.1 CAD
10066-630
Histology Freeze Plate, PolyScience
Specimen Mounting Supplies
The PolyScience Histology Freeze Plate is a fast-freeze cold plate that allows histotechnicians to observe tissue freezing and keep specimen edges flat.

The 877 cm2 (136 in2) freezing surface accommodates multiple specimens simultaneously.

The ultra-cold surface freezes samples quickly, reducing overall processing time by 40% or more.

Working temperature: Fixed at –34.4 °C (–30 °F)

Certifications: cETLus compliant.

Caution: Please note this unit is 240 V, 50 Hz
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR