Hi-Speed Mini Plasmid Kits, IBI Scientific

Supplier: IBI Scientific

IB47101 IB47102
95039-652EA 293.04 CAD
95039-652 95039-654
Hi-Speed Mini Plasmid Kits, IBI Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

IBI's Hi-Speed Mini Plasmid Kit is designed for rapid isolation of plasmid or cosmid DNA from bacterial cultures


  • Sample Size: 1 – 4 ml bacterial culture
  • Expectant Yield: 20-30 µg for high-copy plasmids/3-10µg for low-copy plasmid
  • Operation Time: 30min or less
  • Binding Capacity: up to 30 µg


The purified plasmid DNA is ready to use for restriction enzyme digestion, ligation, PCR, or sequencing reactions. This entire procedure can be completed in under 30 minutes.


The modified alkaline lysis method and Rnase treatment are used to create cleared cell lysate with minimal genomic DNA and RNA contaminants. In the presence of a chaotropic salt, the plasmid DNA in the lysate, binds to the glass-fiber matrix in the spin column. The contaminants are washed off with an ethanol based wash buffer, and the purified plasmid DNA is eluted by a low salt elution buffer or water. This procedure does not require DNA phenol extraction or alcohol precipitation. Typical yields are 20-30µg for high-copy numbered plasmid or 3-10µg for low-copy numbered plasmid.

Order Now

SPECIFICATIONS

Description Hi-Speed Mini Plasmid Kit
Environmentally Preferable
Application
Target molecule Plasmid
Refine target molecule Plasmid
Sample type Bacteria
Method / Format Column (silica-based)
Prep size Mini
Yield 20-30 µg for High Copy Plasmid/ 3-10 µg for Low Copy Plasmid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR