Nalgene® Diagnostic Bottles, HDPE, with Closures, Non-Sterile, Tray Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
322002-9125 322004-9125 322004-9025 322002-9050 322004-9050 322002-9025
16059-674CS 1334.78 CAD
16059-674 16059-678 16059-688 16059-694 16059-696 CA10098-580
Nalgene® Diagnostic Bottles, HDPE, with Closures, Non-Sterile, Tray Pack, Thermo Scientific
Bottles Plastic Bottles
Perform many small volume applications with safe and leakproof Thermo Scientific™ Nalgene™ HDPE diagnostic bottles with closures.
  • Available in natural and amber
These bottles are ideal for use in test kits, sample storage and shipping of small volume, valuable reagents.
Certifications: Amber bottles meet the requirements of light-resistant containers per the current edition of USP.
Packaging: They come packed in rigid trays for easy handling.