Corning 384-well High Content Screening Microplates with Glass Bottom
Supplier: Corning
4581 4583 4585 4587
30621-098CS 501.41 CAD
30621-098 89511-322 89511-326 89511-330
Corning 384-well High Content Screening Microplates with Glass Bottom
Microplates Cell Culture Plates
High optical quality, glass bottom black microplates are ideal for performing high-content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.
  • High optical quality and scratch resistant glass
  • Glass bottom thickness of 200 µm is well suited for imaging microscopy
  • Bottom flatness <50 µm to ensure planarity for imaging devices
  • Low background fluorescence and minimal crosstalk provide the highest possible optical quality for cell-based assays
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk ISO 14001 - Bedford (reg. no. 14.4942)