Thermo Scientific Cryogenics Locator® 7 with Autofill, Thermo Scientific
Supplier: Thermo Fisher Scientific
AY509X32 AY509X33 CS509X24
24269-018CS 1042.34 CAD
24269-018 24269-020 24269-030 24269-083
Thermo Scientific Cryogenics Locator® 7 with Autofill, Thermo Scientific
Dewars and Cryogenic Vessels
LOCATOR 7 Autofill cryogenic storage vessel, designed to attach to your liquid nitrogen tank for automatic filling of aluminum cryogenic vessels with liquid nitrogen for vapor phase storage. The autofill controller minimizes manual checking and refilling vessels with liquid nitrogen on a daily basis, which is critically important for vapor phase storage.
  • Automatically monitors and refills vessels with liquid nitrogen
  • Minimizes manual rechecking and recharging liquid nitrogen vessels on a daily basis
  • Autofill controller features audible and visual low/high
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable