Sample Tubes with External Threads, Simport Scientific
Supplier: Simport Scientific
T501-5T T501-3AT T501-5AT T501-2TPR T501-2ATPR T501-3ATPR T501-4ATPR T501-5ATPR T501-4T T501-2T T501-2AT T501-4AT T501-1ATPR T501-5TPR T501-4TPR T501-4T T501-4TPR T501-5T
CA11006-402CS 299.95 CAD
CA11006-402 CA60872-338 CA60872-342 CA60872-346 CA60872-348 CA60872-350 CA60872-352 CA60872-354 CA60872-433 CA60872-334 CA60872-336 CA60872-340 CA60872-344 CA89135-598 CA11027-092 490010-468 10154-264 76299-336
Sample Tubes with External Threads, Simport Scientific
Tubes Test Tubes
Vial has external threads, providing a smooth and uniform inner surface, thus reducing the risk of contamination.
  • Manufactured from non-toxic polypropylene
  • Height of tube is with cap
  • Caps available with or without silicone washer
  • Caps feature a long skirt and super-fast thread design that allows for single turn seal or removal
  • Tube and cap are made of polypropylene, providing the same coefficient of expansion
Designed for the storage and transportation of biological material. The tube provides strength and clarity and exhibits some unique designMore Product Information