MBP 101 to 1000 µl Graduated Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
112-G 112-GN 112-R100GS 112-RGN
89404-814CS 1030.31 CAD
89404-814 89404-816 490014-508 490014-528
MBP 101 to 1000 µl Graduated Pipette Tips, Thermo Scientific
Pipette Tips
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product.
Autoclavable Tips and Racks: pipette tips and racks are autoclavable at a recommended temperature of 120 °C at 15 pounds of pressure for 15 minutes. Graduated Tips: Provides reference lines to allow a quick check of pipetting procedures by indicating nominal liquid volume in the tip. For use with but not limited to: Pipetman®, Costar®, Eppendorf®, Finnpipette®, Nichiryo, Oxford-Benchmate®, Rainin EDP2™, SMI Airpettor, Socorex®, Titertek®, Volac, and GeneMate®.
A mold release agent is NOT used in the manufacturing of these pipette tips. Metal-free Tips: All materials used in the manufacture of these tips are free of trace metals. A trace metal certificate is available upon request. Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non-sterilized product is not certified as pyrogen-free.) Certificate of analysis is available upon request. Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation. Certificate of irradiation available upon request.
Packaging: Stack Rack comes with 2 stacks of 5 trays of 100 tips each.