MBP 1 to 200 µl Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
110-11 110-96R 110-N H-110-96RNS 110-96RN 110-96RS HRI-110-N
89404-794CS 635.06 CAD
89404-794 89404-796 89404-802 490014-512 490014-516 490014-520 490016-318
MBP 1 to 200 µl Pipette Tips, Thermo Scientific
Pipette Tips
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product.
  • Metal-free Tips: All materials used in the manufacture of these tips are free of trace metals
  • A mold release agent is NOT used in the manufacturing of these pipette tips
Autoclavable Tips & Racks: pipette tips and racks are autoclavable at a recommended temperature of 120°C at 15 pounds of pressure for 15 minutes. For use with but not limitedMore Product Information