X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside), Ultrapure

Supplier: TEKNOVA CA

X1205 X1201
CA100218-934EA 1796.78 CAD
CA100218-934 CA100219-246
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside), Ultrapure
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

Used in conjunction with cloning systems that use b-galactosidase activity as an indicator of nonrecombination.


White powder

Formula: C₁₄H₁₅BrClNO₆
MDL Number: MFCD00005666
CAS Number: 7240-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR