IPTG (isopropyl-β-D-thiogalactopyranoside), Ultrapure

Supplier: TEKNOVA CA

I3305 I3301
CA100218-372EA 619.85 CAD
CA100218-372 CA100219-244 CA100219-816
IPTG (isopropyl-β-D-thiogalactopyranoside), Ultrapure
IPTG (Isopropyl-β-D-thiogalactopyranoside)

IPTG, a non-metabolizable galactose analog, is commonly used in cloning procedures that require induction of β-galactosidase activity


IPTG powder can be stored at Room Temperature. For optimal shelf-life, we recommend storage, with dessicant, at -20oC. IPTG stock solution is typically made at a 1M concentration and stored at -20oC. We recommend making single-use aliquots to prevent repeated freeze-thaw cycles of IPTG in solution.


IPTG is used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies that induce expression of the lac operon in Escherichia coli. IPTG functions by binding to the lacI repressor and altering its conformation, which prevents the repression of the β-galactosidase coding gene lacZ. Teknova IPTG is Dioxane-Free

Formula: C₉H₁₈O₅S
MDL Number: MFCD00063273
CAS Number: 367-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR