D-(+)-Glucose, anhydrous, Ultrapure

Supplier: TEKNOVA CA

Synonyms: Corn sugarD-GlucoseD-(+)-DextroseGlucolinᴅ-(+)-Glucose(2R,3S,4R,5R)-2,3,4,5,6-PentahydroxyhexanalGlucoseDextrose

G5802 G5802
CA100219-258EA 375.87 CAD
CA100219-258 100219-258
D-(+)-Glucose, anhydrous, Ultrapure
D(+)-Glucose

Synonyms: Dextrose, corn sugar, blood sugar.
Description: Crystals.
Applications: Carbon source for the maintenance and growth of microorganisms
Molecular Formula: C6H12O6.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
MDL Number: MFCD00148912
CAS Number: 50-99-7
https://ca.vwr.com/stibo/web/std.lang.all/21/22/2302122.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR