Teknova YEPD Broth
Supplier: TEKNOVA CA
Y9105 Y9110
CA100216-850EA 500.2 CAD
CA100216-850 CA100217-510 CA100218-392
Teknova YEPD Broth
Media for Microbiology Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving.