Teknova YT Broth
Supplier: TEKNOVA CA
Y0110
Y0105
CA100216-678EA
690.9
CAD
CA100216-678
CA100219-880
Teknova YT Broth
Media for Microbiology
Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving.