Teknova SOB Agar
Supplier: Teknova
S0220 S0220
CA100220-032EA 579.43 CAD
CA100220-032 100220-032
Teknova SOB Agar
Media for Microbiology Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving.