Teknova NZYM Broth
Supplier: TEKNOVA CA
N2062
N2063
CA100216-982EA
813.84
CAD
CA100216-982
CA100219-850
Teknova NZYM Broth
Media for Microbiology
Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving.