Tryptone Broth, Teknova
Supplier: TEKNOVA CA
T9205 T9210
CA100217-042EA 392.33 CAD
CA100217-042 CA100218-340
Tryptone Broth, Teknova
Media for Microbiology Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving.