Teknova LB Media, No Salt
Supplier: TEKNOVA CA
L9170
L9165
CA100218-846EA
743.33
CAD
CA100218-846
CA100219-110
Teknova LB Media, No Salt
Media for Microbiology
Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving.