Teknova LB Medium, Low Salt
Supplier: Teknova
L9195 L9190 L9199 L9199 L9195 L9190
CA100219-284EA 848.74 CAD
CA100219-284 CA100217-038 100217-038 100219-284 100217-714 CA100217-714
Teknova LB Medium, Low Salt
Media for Microbiology Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving. Lennox.