Teknova Complete Minimal Dropout Plates Glucose with Three Dropouts
Supplier: TEKNOVA CA
C3460 C3480 C3500 C3461
CA100216-862PK 487.1 CAD
CA100216-862 CA100216-868 CA100217-572 CA100217-576 CA100217-578 CA100217-858
Teknova Complete Minimal Dropout Plates Glucose with Three Dropouts
Media for Microbiology Prepared Media for Microbiology
Plates are shelf-life dated and guaranteed for 4 weeks from the date of shipment
All plates must be stored at 4°C and in the absence of continuous exposure to light . With proper storage, most antibiotic plates are useable for up to 2 months. All plates are sold in sleeves of 20. Complete Minimal Dropout Plates (CM)Also known as Synthictic Dropout Plates (SD).