Teknova Complete Minimal Dropout Plates Glucose with Two Dropouts
Supplier: Teknova
C3220 C3260 C3160 C3180 C3240 C3220 C3220 C3260 C3221 C3200 C3221 C3160 C3200 C3180 C3240 C3261 C3261 C3260
CA100217-848PK 457.33 CAD
100217-564 100217-552 100217-562 100217-550 CA100217-848 CA100217-548 77443-452 CA100217-556 CA100217-854 CA100217-550 CA100217-562 CA100217-552 CA100217-564 77528-950 100217-848 100217-854 100217-556 100217-548
Teknova Complete Minimal Dropout Plates Glucose with Two Dropouts
Media for Microbiology Prepared Media for Microbiology
Plates are shelf-life dated and guaranteed for 4 weeks from the date of shipment
All plates must be stored at 4°C and in the absence of continuous exposure to light . With proper storage, most antibiotic plates are useable for up to 2 months. All plates are sold in sleeves of 20. Complete Minimal Dropout Plates (CM)Also known as Synthictic Dropout Plates (SD).