Teknova Growth Media and Solutions for Manipulating Bacteriophage Lambda, Teknova
Supplier: TEKNOVA CA
M1260
M1200
Y4220
M1201
Y4221
Y4010
Y4210
M1261
CA100216-346PK
782.94
CAD
CA100216-346
CA100216-636
CA100216-658
CA100216-916
CA100217-442
CA100219-022
CA100219-192
CA100219-360
CA100219-564
CA100219-782
Teknova Growth Media and Solutions for Manipulating Bacteriophage Lambda, Teknova
Media for Microbiology
Prepared Media for Microbiology
With proper storage, most antibiotic plates are useable for up to two months.
- Shelf-life dated
- Useable for up to two months
Plates are shelf-life dated and guaranteed for 4 weeks from the date of shipment. All plates must be stored at 4°C and in the absence of continuous exposure to light. With proper storage, most antibiotic plates are useable for up to 2 months. All plates sold in sleeves of 20.