LB Agar Ampicillin, Kanamycin
Supplier: TEKNOVA CA
L1206 L1913
CA100217-336PK 440.73 CAD
CA100217-336 CA100217-338 CA100219-418 CA100219-420 CA100220-080 CA101320-132
LB Agar Ampicillin, Kanamycin
Media for Microbiology Prepared Media for Microbiology
TEKnova's pre-poured plates will save you time. No more media preparation, autoclaving, pouring and the aggravation of doing it over and over again. LB (Luria-Bertani) plates contain 1% tryptone, 0.5% yeast extract, 1.0% NaCl, and 1.5% agar unless otherwise indicated. LB medium is adjusted to a final pH of 7.0 unless otherwise indicated. All plates are tested with at least 4 different recombinant strains of E. coli to demonstrate the efficacy of the antibiotics and colorimetric indicators contained within.
  • Plates are Shelf-Life Dated and Guaranteed for 4 Weeks From the Date of Shipment
  • Must be Stored at 4°C and in the Absence of Continuous Exposure to Light
  • With Proper Storage, Most Antibiotic Plates are Useable for Up to 2 Months
  • All Plates are Sold in Sleeves of 20