LB Agar Ampicillin
Supplier: TEKNOVA CA
L3900 L5902 L3002 L3004 L7202 L7204
CA100217-628PK 1193.06 CAD
CA100217-628 CA100217-632 CA100217-668 CA100217-672 CA100218-186 CA100218-190 CA100220-122 CA100220-124
LB Agar Ampicillin
Media for Microbiology Prepared Media for Microbiology
All plates are tested with at least 4 different recombinant strains of E
  • Plates Shelf-Life Dated and Guaranteed for 4 Weeks From Date of Shipment
  • Must be Stored at 4°C and in Absence of Continuous Exposure to Light
  • With Proper Storage, Most Antibiotic Plates are Useable for Up to 2 Months
  • All Plates Sold in Sleeves of 20 Unless Noted Otherwise
Luria-Bertani plates contain 1% tryptone, 0.5% yeast extract, 1.0% NaCl, and 1.5% agar unless otherwise indicated. LB medium is adjusted to a final pH of 7.0 unless otherwise indicated. All plates are tested with at least 4 different recombinant strains of E. coli to demonstrate the efficacy of the antibiotics and colorimetric indicators contained within.
coli to demonstrate the efficacy of the antibiotics and colorimetric indicators contained within.
Order Now
SPECIFICATIONS
Environmentally Preferable