MBP DNA AWAY™
Supplier: Molecular BioProducts
7010
53509-506CS 1217.31 CAD
53509-506
MBP DNA AWAY™
Cleaning Agents Decontaminants
RNase AWAY® eliminates RNase and DNA from laboratory surfaces
  • Easily Remove RNase Contaminants
  • Safe to Use on Pipettors, Glassware, Plasticware, and Most Metal Surfaces
  • Space-Saving, Convenient Packaging
  • Eliminates Over-Spraying of Decontaminants on Sensitive Equipment
The surface decontaminant is ideal for decontaminating apparatus, benchtops, glassware, and plasticware. The decontaminant leaves no residue to interfere with gel polymerization or staining and contains no strong acids. DNA AWAY™ decontaminant is ideal for removing DNase and DNA contamination.
Certifications: Biotechnology grade. Application Conformity Pass. Store frozen. Irritant. Hazard Statements: H315 / H319 / H335. Precautionary Statements: P280 / P302+P352 / P304+P340 / P305+P351+P338.