Miniature Stopcock, Dynalon
Supplier: Dynalon
228285 228275 228265 228255
71070-124CS 79.41 CAD
71070-124 71070-126 71070-128 71070-130
Miniature Stopcock, Dynalon
Stopcocks
Miniature stockpocks allow for easy turning of key which requires no lubricant, thus eliminating blockage of the bores with grease.