Tubing Connectors, Polypropylene, Dynalon
Supplier: Dynalon
222595 222955 227215
62849-331CS 904.93 CAD
62849-331 62849-345 62849-374 62849-310 62849-326 62849-350 62849-380
Tubing Connectors, Polypropylene, Dynalon
Fittings and Connectors
Excellent chemical resistance, autoclavable polypropylene
Features a serrated connecting surfaces to prevent tubing slipping and withstands flows to 20 p.s.i.
Ordering information: Elasticity of tubing will influence choice of size. Dimensions listed are nominal.