Water, PCR grade
Supplier: IBI Scientific
Synonyms:
Hydrogen oxide
IBI RT-PCR Certified and Nuclease-Free Water is ideal for all applications in a molecular biology lab including PCR, RT-PCR, restriction enzyme assays, modifying enzyme assays, transfection, cloning, transformation, and all general molecular biology lab procedures.
- Bottled in 2 ml vials
- 0.1 µm sterile-filtered into certified-clean polypropylene vials
- Steam-sterilized (autoclaved)
- 18.2 Megohm deionized
IBI water is manufactured under stringent conditions. The purification process includes continuous deionization, reverse osmosis, UV-treatment, 0.2 µm filtration, followed by steam sterilization in an autoclave.
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 Melting Pt: 0 Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 Merck Index: 13,10098 |
Specification Test Results
Cations & Anions Free | Yes |
Mycoplasma Free | Yes |
Sterility Assurance By | 0.1 µm pore filter |
Resistivity | 18.2 mOhms |
Endotoxin (LAL) | ≤ 0.1 EU/ml |
Electrophoresis | Yes |
PCR | Yes |
RT-PCR | Yes |
MEA Approved | N/A |
RNAse | None Detected |
DNAse | None Detected |
Nickase | None Detected |
Protease | None Detected |
Phosphatase | None Detected |
Mycoplasma | None Detected |
Iron (Fe) | Not Tested |
Sterility Assured | 0.1 µm sterile-filtered and 2 X autoclaved |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...