Water, Molecular Biology Grade
Supplier: IBI Scientific
Synonyms:
Hydrogen oxide
IBI Molecular Biology Grade, Nuclease-Free Water is ideal for all applications in a molecular biology lab including PCR, RT-PCR, restriction enzyme assays, modifying enzyme assays, transfection, cloning, transformation and all general molecular biology lab procedures.
IBI Molecular Biology Grade Water is manufactured under stringent conditions. The purification process for this product includes continuous deionization, reverse osmosis, UV-treatment, 0.2 µm filtration, followed by steam sterilization in an autoclave.
Formula:
H₂O MW: 18.02 g/mol |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Specification Test Results
Cations & Anions Free | Yes |
Mycoplasma Free | Yes |
Sterility Assurance By | 0.1 µm pore filter |
Resistivity | 18.2 mOhms |
Endotoxin (LAL) | ≤ 0.1 EU/ml |
Electrophoresis | Yes |
PCR | Yes |
RT-PCR | Yes |
MEA Approved | N/A |
RNAse | None Detected |
DNAse | None Detected |
Nickase | None Detected |
Protease | None Detected |
Phosphatase | None Detected |
Mycoplasma | None Detected |
Heavy Metals | Not Tested |
Iron (Fe) | Not Tested |
Sterility Assured | 0.1 µm sterile-filtered and 2 X autoclaved |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...