Water, Cell Culture Grade

Supplier: IBI Scientific

IB42010 IB42011 IB42012 IB42020 IB42021 IB42030
10065-936EA 87.15 CAD
10065-936 10065-938 10065-940 10065-942 10065-944 10065-946
Water, Cell Culture Grade
Water

IBI Cell Culture Grade, Pyrogen/Endotoxin Free Water is prepared for nuclear transfer, transfection, transformation, cell and tissue culture, LAL testing, and all other aqueous solution preparations that require stringent quality control.


  • Certified endotoxin-free and pyrogen-free
  • 0.1 µm sterile-filtered into certified-clean polycarbonate bottles
  • Double distilled and steam-sterilized


IBI Cell Culture Grade Water is tested for use in critical assays and applications. The water is endotoxin-free (to the maximum detection limit of ˂0.005 EU/mL by turbidimetric assay).
The purification process of our Cell Culture Grade Water includes distillation, continuous deionization, reverse osmosis, 0.1 µm filtration, followed by steam sterilization in an autoclave.

Formula: H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Cations & Anions Free Yes
Mycoplasma Free Yes
Sterility Assurance By 0.1 µm pore filter
Resistivity 18.2 mOhms
Endotoxin (LAL) ≤ 0.005 EU/ml
Electrophoresis Yes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR