Accucore™ Phenyl-Hexyl Columns, Thermo Scientific
Supplier: Thermo Scientific
17926-154630 17926-153030 17926-152130 17926-104630 17926-103030 17926-102130 17926-054630 17926-052130 17926-053030 17926-014005 17926-012105 17926-013005
10038-704EA 2208.49 CAD
10038-704 10038-706 10038-708 10038-710 10038-712 10038-714 10038-716 10038-740 10038-742 10039-096 10039-098 10039-100 10039-102
Accucore™ Phenyl-Hexyl Columns, Thermo Scientific
Chromatography Columns
Separate aromatic and moderately polar compounds with improved speed and resolution using Thermo Scientific™ Accucore Phenyl-Hexyl columns.
Rugged 2.6 µm solid core particles enable fast, high efficiency separations at low backpressures. The phenyl-hexyl stationary phase offers a mixed-mode separation mechanism: the C6 chain provides hydrophobic retention while the phenyl ring enhances pi-pi interactions. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Ordering information: Guard cartridges require use ofMore Product Information