Accucore™ C8 Columns, Thermo Scientific
Supplier: Thermo Scientific
17226-154630 17226-153030 17226-152130 17226-104630 17226-103030 17226-102130 17226-052130 17226-033030 17226-032130 17226-013005 17226-014005 17226-012105
10038-812EA 2219.74 CAD
10038-812 10038-814 10038-816 10038-818 10038-820 10038-822 10038-824 10038-826 10038-828 10038-830 10038-834 10038-836 10038-838 10038-840
Accucore™ C8 Columns, Thermo Scientific
Chromatography Columns
Improve separation speed and resolution while maintaining low backpressures using Thermo Scientific™ Accucore C8 columns.
Rugged 2.6 µm solid core particles ensure high efficiencies and enable compatibility with both HPLC and UHPLC platforms. The high coverage C8 bonded phase provides optimal retention of analytes with moderate hydrophobicity. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).