Accucore™ RP-MS Columns, Thermo Scientific
Supplier: Thermo Scientific
17626-153030 17626-154630 17626-152130 17626-103030 17626-104630 17626-054630 17626-102130 17626-053030 17626-052130 17626-032130 17626-014005 17626-013005 17626-012105
10039-104EA 2177.23 CAD
10039-104 10039-106 10039-108 10039-110 10039-112 10039-114 10039-116 10039-118 10039-120 10039-126 10039-128 10039-130 10039-134
Accucore™ RP-MS Columns, Thermo Scientific
Chromatography Columns
Optimize LC/MS analyses with Thermo Scientific™ Accucore RP-MS columns.
Rugged 2.6 µm solid-core particles enable fast, high-efficiency separations at low backpressures. The stationary phase provides slightly lower retention than C18 and has more effective surface coverage for improved peak efficiencies. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (1More Product Information