VWR® 99% Protein-Free Amber Tubing
Supplier: VWR International
62996-380 62996-695
62996-380PK 144.44 CAD
62996-380 62996-695
VWR® 99% Protein-Free Amber Tubing
Tubing Transfer Tubing
Extensively leached, washed and surface treated to undetectable protein levels.
  • 99% protein-free
  • Amber color
Compatible with steam, ETO, or gamma radiation.