Blades For #3 Scalpel
Supplier: C & A Scientific
9310 9311 9312 9315
470094-362BX 62.25 CAD
470094-362 470094-382 470094-388 470094-460
Blades For #3 Scalpel
Scalpels and Scalpel Assemblies Scalpel Blades
High Quality Blades For Fine Dissection
  • Individually Wrapped In Foil
  • Available In Packs Or Boxes
  • Multiple Shapes Available
Scalpel blades to fit number 3 handles. Each blade is individually wrapped.