MAbPac™ SCX-10 Columns 5 µm, Thermo Scientific
Supplier: Thermo Scientific
088783 078655 078656 085198
10044-354EA 9709.34 CAD
10044-354 10044-356 10044-844 10044-846 10044-922
MAbPac™ SCX-10 Columns 5 µm, Thermo Scientific
Chromatography Columns
For high-resolution, high efficiency analysis of monoclonal antibodies (MAbs) and associated variants, use Thermo Scientific™ MAbPac SCX-10 strong cation-exchange columns
The unique nonporous pellicular resin provides the resolving power to separate MAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.