ProPac™ WCX-10 Cation Exchange Protein Columns, Thermo Scientific
Supplier: Thermo Scientific
074600 088766 088767 088768 088778 088779 054993 054994 063472 063474 063480
10042-822EA 2676.9 CAD
10042-822 10044-322 10044-324 10044-326 10044-346 10044-348 10044-380 10044-382 10044-508 10044-510 10044-512
ProPac™ WCX-10 Cation Exchange Protein Columns, Thermo Scientific
Chromatography Columns
For separations of proteins and their variants using Thermo Scientific™ ProPac SCX and WCX strong and weak cation-exchange columns, which provide exceptionally high resolution.
  • Characterisation and quality control assessment of monoclonal antibodies and other proteins
  • Useful for characterisation of related protein variants including deamidation and monoclonal antibodies lysine truncation variants
Based on stationary phases composed of 10 µm nonporous polymeric beads, these columns resolve isoforms that differ by a single charged residue. A thin,More Product Information