Acclaim™ Trinity Q1 LC Columns, Thermo Scientific
Supplier: Thermo Scientific
079719 079717 079715 083242 083241 083244
10045-856EA 502.78 CAD
10045-856 10148-466 10148-490 10148-492 10148-494 10148-496
Acclaim™ Trinity Q1 LC Columns, Thermo Scientific
Chromatography Columns
Simultaneously separate the herbicides diquat and paraquat with Thermo Scientific™ Acclaim Trinity Q1 high-efficiency LC columns.
These tri-mode (WCX, WAX, RP) columns offer unmatched high-resolution and high-throughput trace analysis through LC-MS/MS and LC-UV methods. The stationary phase, based on Nanopolymer Silica Hybrid (NSH) technology, provides multiple retention mechanisms, including reversed-phase, anion-exchange, and cation-exchange, for maximum flexibility in method development. Easily optimize the selectivity to accelerate chromatographic separations and increase productivity.
Ordering information: Guard Cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2 (Part No. 10046-750).