Syncronis™ C8 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
97202-052130 97202-102130 97203-104630 97203-154630 97205-014001 97205-104630 97205-154630 97205-253030 97205-254030 97205-254630
10046-656EA 2340.58 CAD
10046-656 10046-662 10046-668 10046-670 10046-672 10046-678 10046-696 10046-698 10046-702 10046-704 10046-708 10046-710 10046-712
Syncronis™ C8 HPLC Columns, Thermo Scientific
Chromatography Columns
Improve the consistency and reliability of your separations using Thermo Scientific™ Syncronis C8 columns.
Highly pure, high surface area silica and dense stationary phase coverage minimize secondary interactions and deliver reproducible, high efficiency separations. Double end-capping provides extra surface coverage and inertness towards basic compounds. Manufactured and rigorously tested in ISO9000 facilities, these columns are designed to meet a range of analytical needs, including fast method development, quantitative analysis, and quality control where less retention is required than when using a C18 column.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).