Syncronis™ C18 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
97102-032130 97102-052130 97102-054630 97102-102130 97103-053030 97103-102130 97103-103030 97103-104630 97103-152130 97103-154630 97105-012101 97105-014001 97105-052130 97105-054630 97105-102130 97105-104630 97105-152130 97105-153030 97105-154030 97105-154630 97105-252130 97105-253030 97105-254030 97105-254630 97105-159070A 97105-159270A 97105-159370A 97105-259070A 97105-259270A 97105-018823A 97105-019023A 97105-019323A 97105-159270XL 97105-259270XL 97105-019323B
10044-058EA 2413.46 CAD
10044-058 10044-060 10044-064 10044-066 10044-068 10044-072 10044-074 10044-076 10044-078 10044-080 10044-082 10044-084 10044-086 10046-610 10046-612 10046-614 10046-624 10046-630 10046-632 10046-634 10046-636 10046-638 10046-640 10046-642 10046-644 10046-646 10046-648 10046-650 10046-652 76332-286 76332-288 76332-290 76332-292 76332-294 76332-296 76332-298 76332-302 76332-418 76332-420 76332-422 76332-424
Syncronis™ C18 HPLC Columns, Thermo Scientific
Chromatography Columns
Achieve repeatable and consistent liquid chromatography (LC) separations using Thermo Scientific™ Syncronis™ C18 columns.
  • Highly reproducible separations
  • Excellent peak shapes for enhanced resolution and sensitivity
  • Dense surface coverage of silica minimizes secondary interactions and peak tailing
  • Double endcapped for extra surface coverage
  • Exceptional column-to-column consistency
  • Manufactured, packed, and tested in ISO-9000-accredited facilities
  • Enhanced automated packing process
Highly pure, high surface area silica and dense stationary phase coverage minimize secondary interactions and ensure robust, high-resolution separations. Double end-capping provides extra surface coverage and inertness toward basic compounds. Manufactured and rigorously tested in ISO 9000 facilities, these columns are designed to meet a range of analytical needs including fast method development, quantitative analysis, and quality control.
Available in three particle sizes - 1.7 μm for rapid ultrahigh-performance LC (UHPLC) separations, and 3 and 5 μm for more traditional HPLC analysis. Rigorously tested stationary phase bonded to the highest quality silica ensures consistent, predictable LC performance. Every column individually tested to meet stringent retention, efficiency, and peak symmetry specifications.