Accucore™ 150-C4 Columns, Thermo Scientific
Supplier: Thermo Scientific
16526-154630 16526-157569 16526-152130 16526-104630 16526-102130 16526-103030 16526-052130 16526-014005 16526-032130 16526-013005 16526-012105
10038-904EA 3387.61 CAD
10038-904 10038-906 10038-908 10038-910 10039-156 10039-158 10039-160 10039-162 10039-166 10039-170 10039-172 10039-174 10039-176 10039-178 10039-180
Accucore™ 150-C4 Columns, Thermo Scientific
Chromatography Columns
The 150A pore diameter solid core particles used in Thermo Scientific Accucore 150-C4 columns are designed specifically to provide the optimum combination of retention and resolution for proteins and larger peptides.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).