Hypersil™ ODS-2 C18 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
31603-054630 31603-104630 31603-154630 31605-014001 31605-054630 31605-104630 31605-124030 31605-124630 31605-154630 31605-204630 31605-254030 31605-254630 31603-014001
10042-710EA 1475.41 CAD
10042-710 10042-714 10042-716 10042-730 10042-736 10042-744 10042-750 10042-754 10042-756 10042-760 10042-764 10042-766 10042-772 10042-774 10042-776 10046-158
Hypersil™ ODS-2 C18 HPLC Columns, Thermo Scientific
Chromatography Columns
Classical Hypersil stationary phases have global recognition as an industry standard in HPLC, providing an effective analytical tool.
Hypersil phases are well established and referenced in many HPLC methods worldwide.
Certifications: Manufactured under ISO 9001-2000 quality guidelines.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).