Hypersil GOLD™ PFP HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
25402-102130-V 25402-152130-V
10041-448EA 1693.73 CAD
10041-448 10041-454 10041-456 10041-458 10041-462 10041-464 10041-466 10041-468 10041-470 10041-474 10041-486 10041-492 10041-494 10041-496 10041-498 10041-500 10041-502 10041-508 10041-510 10041-512 10041-514 10041-516 10041-518 10041-522 10041-524 10041-526 10041-528 10041-530 10041-532 10041-538 10041-540 10041-542 10041-544 10041-546 10041-548 10045-572 10045-574 10045-576 10045-578 10045-580 10045-588 10045-590 10045-604 10045-606 10045-610 10045-616 10045-618 10045-622 10045-624
Hypersil GOLD™ PFP HPLC Columns, Thermo Scientific
Chromatography Columns
Analyze difficult-to-resolve mixtures of halogenated compounds and non-halogenated polar compounds with Thermo Scientific™ Hypersil GOLD™ PFP (pentafluorophenyl) HPLC columns.
These columns provide an alternative selectivity to C18 columns in reversed phase applications with excellent peak shape and sensitivity.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm ID require the 1 mm UNIGUARD Holder (10046-746), 2.1/3 mm ID require the 2/3 mm UNIGUARD Holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD Holder (10046-744)