Reacti-Vap Evaporators, Thermo Scientific
Supplier: Thermo Scientific
TS-18826 TS-18825
10055-298EA 2234.11 CAD
10055-298 10055-300
Reacti-Vap Evaporators, Thermo Scientific
Evaporators Centrifugal Evaporators
Manifolds for easy sample evaporation, with integrated pressure-relief valve to protect against excessive gas flow and dangerous pressure build up
Easy set up: attach to corresponding Reacti-Therm Module, attach tubing from gas supply, and lower into position over samples and start gas flow.
Ordering information: For a list of compatible aluminum alloy blocks, please see Reacti-Block Aluminum Blocks, Thermo Scientific.