SWAP-IT GC MS Interface System, Thermo Scientific
Supplier: Thermo Scientific
290SG021 290SG001 290SG000 290SG004 290SG002 290SG025 290SG020 290SG032 290SG003
10055-976EA 584.94 CAD
10055-976 10056-038 10055-038 10055-034 10055-132 10056-040 10055-036 10055-032 10055-130
SWAP-IT GC MS Interface System, Thermo Scientific
Chromatography Supplies Chromatography Spare Parts
The SWAP-IT GC-MS Interface provides a powerful tool for switching capillary columns without the need to vent the MS
A specially deactivated, robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector. The small ID of the transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection, and finger-tight connections make exchanging the analytical column quick and easy without the need for tools. This system works without the requirement for additional He-purge. (Kits contain Nuts, Ferrules and Accessories in addition to the interface).